Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321829.1 | NP_001308758.1 | 3 | 469 | ||
NM_018017.3 | NP_060487.2 | 2 | 344 | |||
NM_153249.1 | NP_694981.1 | 3 | 469 | |||
NR_135815.1 | 3 | 469 | ||||
XM_011539916.2 | XP_011538218.1 | 2 | 436 | |||
GenBank mRNA | AK092620.1 | - | 3 | 452 | ||
AK307517.1 | - | 2 | 321 | |||
BC020842.1 | - | 2 | 336 | |||
BC070368.1 | - | 2 | 315 | |||
BC100670.1 | - | 3 | 396 | |||
BC103499.1 | - | 2 | 271 | 40001826 | ||
BC105632.1 | - | 2 | 271 | 40001824 | ||
BC107430.1 | - | 2 | 304 | |||
BC152306.1 | - | 2 | 275 | 40012131 | ||
BX537964.1 | - | 2 | 316 | |||
BX643041.1 | - | 2 | 355 |