Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143824.1 | NP_001137296.1 | 10, 11 | 1210 | ||
NM_018018.4 | NP_060488.2 | 11, 12 | 1402 | |||
XM_005268997.2 | XP_005269054.1 | 10, 11 | 1211 | |||
GenBank mRNA | AB055003.1 | - | 9, 10 | 1034 | ||
AF193836.1 | - | 10, 11 | 1190 | |||
AF305814.1 | - | 11, 12 | 1367 | |||
AK291168.1 | - | 10, 11 | 1211 | |||
BC069819.1 | - | 10, 11 | 1263 | 7262399 | ||
BC101827.1 | - | 9, 10 | 1047 | 8069333 | ||
BC104913.1 | - | 9, 10 | 1046 | 8143916 |