Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143824.1 | NP_001137296.1 | 14 | 1561 | ||
NM_018018.4 | NP_060488.2 | 15 | 1753 | |||
XM_005268997.2 | XP_005269054.1 | 14 | 1562 | |||
GenBank mRNA | AB055003.1 | - | 13 | 1385 | ||
AF193836.1 | - | 14 | 1541 | |||
AF305814.1 | - | 15 | 1718 | |||
AK291168.1 | - | 14 | 1562 | |||
BC069819.1 | - | 14 | 1614 | 7262399 | ||
BC101827.1 | - | 13 | 1398 | 8069333 | ||
BC104913.1 | - | 13 | 1397 | 8143916 |