Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143824.1 | NP_001137296.1 | 16 | 2277 | ||
NM_018018.4 | NP_060488.2 | 17 | 2469 | |||
XM_005268997.2 | XP_005269054.1 | 16 | 2278 | |||
GenBank mRNA | AB055003.1 | - | 15 | 2101 | ||
AF305814.1 | - | 17 | 2434 | |||
AK291168.1 | - | 16 | 2278 |