Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006634.2 | NP_001006635.1 | 8 | 711 | ||
NM_018054.5 | NP_060524.4 | 8 | 711 | |||
XM_011545873.2 | XP_011544175.1 | 8 | 704 | |||
XM_011545874.2 | XP_011544176.1 | 9 | 786 | |||
XM_011545875.2 | XP_011544177.1 | 8 | 704 | |||
XM_011545876.2 | XP_011544178.1 | 8 | 704 | |||
XM_011545877.2 | XP_011544179.1 | 8 | 642 | |||
XM_011545878.2 | XP_011544180.1 | 8 | 704 | |||
XM_017023390.1 | XP_016878879.1 | 8 | 704 | |||
GenBank mRNA | AJ306731.1 | - | 8 | 594 | ||
AJ306732.1 | - | 8 | 594 | |||
AK023281.1 | - | 5 | 363 | |||
AK027567.1 | - | 8 | 664 | |||
AK291691.1 | - | 8 | 664 | |||
AK301500.1 | - | 6 | 400 | |||
BC080195.1 | - | 8 | 664 | 6172310 | ||
BU187683.1 | - | 8 | 654 |