Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304496.1 | NP_001291425.1 | 6 | 1464 | ||
NM_018073.7 | NP_060543.5 | 7 | 1947 | |||
GenBank mRNA | AF360739.1 | - | 7 | 1900 | ||
AK001231.1 | - | 4 | 1152 | |||
AK022923.1 | - | 4 | 1117 | |||
AK056523.1 | - | 7 | 1909 | |||
AK291166.1 | - | 7 | 1859 | |||
AK301120.1 | - | 5 | 1179 | |||
AL832730.1 | - | 7 | 1897 | |||
BC109063.2 | - | 7 | 1866 | 40033795 |