Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256875.1 | NP_001243804.1 | 5 | 652 | ||
NM_018101.3 | NP_060571.1 | 6 | 555 | |||
GenBank mRNA | AK001330.1 | - | 6 | 475 | ||
AK022104.1 | - | 5 | 557 | |||
AK022606.1 | - | 5 | 461 | |||
AK222549.1 | - | 6 | 475 | |||
AK222568.1 | - | 6 | 429 | |||
AK291917.1 | - | 5 | 507 | |||
AK302116.1 | - | 4 | 319 | |||
AY508815.2 | - | 6 | 475 | |||
BC000703.2 | - | 5 | 548 | 3347875 | ||
BC001651.2 | - | 5 | 566 | 2822261 | ||
BC008079.2 | - | 6 | 408 | 2959649 | ||
BC016944.1 | - | 6 | 428 | 3457122 |