Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 3 | 320 | ||
NM_152879.2 | NP_690618.2 | 4 | 385 | |||
XM_005246107.3 | XP_005246164.1 | 2 | 52 | |||
XM_005246108.1 | XP_005246165.1 | 2 | 206 | |||
XM_011512028.2 | XP_011510330.1 | 4 | 1844 | |||
XM_011512029.2 | XP_011510331.1 | 4 | 1845 | |||
XM_011512030.2 | XP_011510332.1 | 4 | 441 | |||
XM_011512031.2 | XP_011510333.1 | 4 | 315 | |||
XM_011512032.2 | XP_011510334.1 | 4 | 299 | |||
XM_011512033.2 | XP_011510335.1 | 4 | 1846 | |||
XM_011512035.1 | XP_011510337.1 | 2 | 186 | |||
XM_011512037.2 | XP_011510339.1 | 2 | 290 | |||
XM_011512039.2 | XP_011510341.1 | 4 | 1846 | |||
XM_011512040.2 | XP_011510342.1 | 2 | 222 | |||
XM_017005138.1 | XP_016860627.1 | 4 | 723 | |||
XM_017005139.1 | XP_016860628.1 | 2 | 215 | |||
XM_017005140.1 | XP_016860629.1 | 4 | 317 | |||
GenBank mRNA | AB078966.1 | - | 4 | 385 | ||
AK308846.1 | - | 2 | 206 | |||
AK311044.1 | - | 2 | 139 | |||
D63479.3 | - | 4 | 326 | |||
D73409.1 | - | 3 | 321 |