Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 2, 3 | 199 | ||
XM_005249205.1 | XP_005249262.1 | 2, 3 | 260 | |||
GenBank mRNA | AK293016.1 | - | 2, 3 | 204 | ||
BC016279.1 | - | 2, 3 | 189 | 3908895 | ||
BI768787.1 | - | 2, 3 | 217 |