Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 3, 4 | 254 | ||
XM_005249205.1 | XP_005249262.1 | 3, 4 | 315 | |||
XM_011514723.2 | XP_011513025.2 | 2, 3 | 190 | |||
GenBank mRNA | AK293016.1 | - | 3, 4 | 259 | ||
BC016279.1 | - | 3, 4 | 244 | 3908895 | ||
BI768787.1 | - | 3, 4 | 272 |