Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 9 | 1537 | ||
XM_005249205.1 | XP_005249262.1 | 9 | 1598 | |||
XM_011514723.2 | XP_011513025.2 | 8 | 1473 | |||
GenBank mRNA | AK001407.1 | - | 4 | 1087 |