Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032293.2 | NP_001027464.1 | 11 | 1676 | ||
NM_001098507.1 | NP_001091977.1 | 12 | 1769 | |||
NM_003457.3 | NP_003448.1 | 11 | 1721 | |||
XM_011525185.2 | XP_011523487.1 | 12 | 1901 | |||
XM_017025020.1 | XP_016880509.1 | 10 | 1649 | |||
GenBank mRNA | AF046001.1 | - | 11 | 1775 | ||
AK226172.1 | - | 12 | 1742 | |||
AK291801.1 | - | 11 | 1684 | |||
BC002372.2 | - | 11 | 1676 | 2822814 | ||
BC008023.1 | - | 12 | 1435 | 3538197 |