Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167856.1 | NP_001161328.1 | 16 | 2285 | ||
NM_018183.3 | NP_060653.3 | 16 | 2282 | |||
XM_005253572.3 | XP_005253629.1 | 17 | 2423 | |||
XM_005253573.4 | XP_005253630.1 | 18 | 2833 | |||
XM_005253575.3 | XP_005253632.1 | 17 | 2420 | |||
XM_005253576.3 | XP_005253633.1 | 18 | 2464 | |||
XM_006719473.3 | XP_006719536.1 | 19 | 2589 | |||
XM_006719474.3 | XP_006719537.1 | 18 | 2542 | |||
XM_011538533.2 | XP_011536835.1 | 19 | 2583 | |||
XM_011538534.2 | XP_011536836.1 | 16 | 2321 | |||
XM_011538535.2 | XP_011536837.1 | 16 | 2323 | |||
XM_017019556.1 | XP_016875045.1 | 17 | 2407 | |||
XM_017019557.1 | XP_016875046.1 | 17 | 2414 | |||
XM_017019558.1 | XP_016875047.1 | 18 | 2477 | |||
XM_017019559.1 | XP_016875048.1 | 17 | 2544 | |||
GenBank mRNA | AB014772.1 | - | 16 | 2282 | ||
AK001695.1 | - | 17 | 2420 | |||
AK122703.1 | - | 18 | 2838 | |||
AK307488.1 | - | 16 | 2321 | |||
BC030544.1 | - | 17 | 2416 | |||
BC133704.1 | - | 16 | 2285 | 40124902 |