Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018191.3 | NP_060661.3 | 13 | 3845 | ||
XM_005266441.2 | XP_005266498.1 | 12 | 3765 | |||
XM_011535133.1 | XP_011533435.1 | 13 | 3871 | |||
XM_011535134.1 | XP_011533436.1 | 13 | 3771 | |||
XM_011535135.2 | XP_011533437.1 | 13 | 3729 | |||
GenBank mRNA | AB209222.1 | - | 4 | 3974 | ||
AF334406.1 | - | 13 | 3820 | |||
AJ319660.1 | - | 13 | 3792 | |||
AK001578.1 | - | 5 | 2722 | |||
AK090807.1 | - | 7 | 2965 | |||
AK096654.1 | - | 13 | 3835 | |||
AL833821.1 | - | 13 | 3755 |