Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193360.1 | NP_001180289.1 | 10 | 3035 | ||
NM_001193361.1 | NP_001180290.1 | 9 | 2951 | |||
NM_001193362.1 | NP_001180291.1 | 10 | 3194 | |||
NM_001193363.1 | NP_001180292.1 | 9 | 3110 | |||
NM_018199.3 | NP_060669.1 | 8 | 2571 | |||
NR_034164.1 | 11 | 3125 | ||||
NR_034165.1 | 11 | 3284 | ||||
XM_005267817.3 | XP_005267874.1 | 8 | 2501 | |||
XM_011536908.2 | XP_011535210.1 | 8 | 2589 | |||
XM_017021421.1 | XP_016876910.1 | 9 | 2634 | |||
GenBank mRNA | AK001600.1 | - | 8 | 2499 | ||
AK056025.1 | - | 11 | 3071 | |||
AL834434.1 | - | 8 | 2493 | |||
AU143711.1 | - | 1 | 66 | |||
BX647767.1 | - | 9 | 2901 | |||
CR936728.1 | - | 10 | 2910 | |||
CR936729.1 | - | 9 | 2918 |