Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 2, 3 | 626 | ||
NM_018212.4 | NP_060682.2 | 2, 3 | 626 | |||
XM_005273187.2 | XP_005273244.1 | 3, 4 | 666 | |||
XM_011544229.2 | XP_011542531.2 | 2, 3 | 1085 | |||
XM_017001746.1 | XP_016857235.1 | 2, 3 | 1832 | |||
XM_017001747.1 | XP_016857236.1 | 2, 3 | 1085 | |||
XM_017001748.1 | XP_016857237.1 | 2, 3 | 1085 | |||
XM_017001749.1 | XP_016857238.1 | 2, 3 | 1085 | |||
XM_017001750.1 | XP_016857239.1 | 2, 3 | 1085 | |||
XM_017001751.1 | XP_016857240.1 | 2, 3 | 974 | |||
XM_017001752.1 | XP_016857241.1 | 2, 3 | 1085 | |||
XM_017001753.1 | XP_016857242.1 | 3, 4 | 636 | |||
XM_017001754.1 | XP_016857243.1 | 2, 3 | 580 | |||
XM_017001755.1 | XP_016857244.1 | 2, 3 | 323 | |||
XM_017001756.1 | XP_016857245.1 | 2, 3 | 323 | |||
XM_017001757.1 | XP_016857246.1 | 2, 3 | 226 | |||
GenBank mRNA | AF519769.1 | - | 2, 3 | 442 | ||
AK290179.1 | - | 2, 3 | 570 | |||
AY345143.1 | - | 2, 3 | 530 | |||
BC065238.1 | - | 1, 2 | 44 | |||
BC095481.1 | - | 2, 3 | 625 | 30348815 | ||
CB961874.1 | - | 2, 3 | 652 | |||
EU255274.1 | - | 1, 2 | 177 | |||
KM214203.1 | - | 1, 2 | 173 | |||
KM214204.1 | - | 1, 2 | 173 |