Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 5 | 1199 | ||
NM_018212.4 | NP_060682.2 | 5 | 1199 | |||
XM_005273187.2 | XP_005273244.1 | 6 | 1239 | |||
XM_011544229.2 | XP_011542531.2 | 6 | 1715 | |||
XM_017001746.1 | XP_016857235.1 | 7 | 2474 | |||
XM_017001747.1 | XP_016857236.1 | 7 | 1727 | |||
XM_017001748.1 | XP_016857237.1 | 5 | 1658 | |||
XM_017001749.1 | XP_016857238.1 | 6 | 1715 | |||
XM_017001750.1 | XP_016857239.1 | 7 | 1727 | |||
XM_017001751.1 | XP_016857240.1 | 5 | 1547 | |||
XM_017001752.1 | XP_016857241.1 | 5 | 1658 | |||
XM_017001753.1 | XP_016857242.1 | 8 | 1278 | |||
XM_017001754.1 | XP_016857243.1 | 7 | 1222 | |||
XM_017001755.1 | XP_016857244.1 | 7 | 965 | |||
XM_017001756.1 | XP_016857245.1 | 5 | 896 | |||
XM_017001757.1 | XP_016857246.1 | 5 | 799 | |||
GenBank mRNA | AF519769.1 | - | 5 | 1015 | ||
AK096246.1 | - | 3 | 495 | |||
AK290179.1 | - | 5 | 1143 | |||
AY345143.1 | - | 5 | 1103 | |||
BC065238.1 | - | 4 | 617 | |||
BC095481.1 | - | 5 | 1198 | 30348815 | ||
EU255274.1 | - | 4 | 750 | |||
KM214203.1 | - | 5 | 803 | |||
KM214204.1 | - | 5 | 803 |