Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161344.1 | NP_001154816.1 | 9 | 1133 | ||
NM_001161345.1 | NP_001154817.1 | 9 | 1133 | |||
NM_001161346.1 | NP_001154818.1 | 9 | 1097 | |||
NM_001161347.1 | NP_001154819.1 | 7 | 857 | |||
NM_018223.2 | NP_060693.2 | 9 | 1010 | |||
GenBank mRNA | AF170724.1 | - | 9 | 1139 | ||
AK001658.1 | - | 9 | 990 | |||
AK027687.1 | - | 9 | 1091 | |||
AK097671.1 | - | 7 | 788 | |||
AK302785.1 | - | 7 | 842 | |||
AK304333.1 | - | 9 | 1120 | |||
BC012072.1 | - | 8 | 1021 | 4650348 |