Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161344.1 | NP_001154816.1 | 17, 18 | 2039 | ||
NM_001161345.1 | NP_001154817.1 | 17, 18 | 2036 | |||
NM_001161346.1 | NP_001154818.1 | 17, 18 | 2003 | |||
NM_001161347.1 | NP_001154819.1 | 15, 16 | 1763 | |||
NM_018223.2 | NP_060693.2 | 17, 18 | 1916 | |||
GenBank mRNA | AF170724.1 | - | 17, 18 | 2045 | ||
AK001658.1 | - | 17, 18 | 1896 | |||
AK027687.1 | - | 17, 18 | 1997 | |||
AK054917.1 | - | 3, 4 | 944 | |||
AK090948.1 | - | 4, 5 | 1012 | |||
AK097671.1 | - | 15, 16 | 1762 | |||
AK302785.1 | - | 15, 16 | 1748 | |||
AK304333.1 | - | 17, 18 | 2023 | |||
AL137561.1 | - | 881 | ||||
BC012072.1 | - | 16, 17 | 1927 | 4650348 |