Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018229.3 | NP_060699.3 | 1, 2 | 511 | ||
XM_006720196.2 | XP_006720259.1 | 1, 2 | 140 | |||
XM_011536940.2 | XP_011535242.1 | 3 | 483 | |||
GenBank mRNA | AK001675.1 | - | 1, 2 | 356 | ||
AK225345.1 | - | 1, 2 | 358 | |||
AK303999.1 | - | 3 | 385 | |||
AL136685.1 | - | 1, 2 | 490 | |||
BC013174.1 | - | 1, 2 | 475 | 3924075 |