Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286349.1 | NP_001273278.1 | 5 | 961 | ||
NM_018240.6 | NP_060710.3 | 7 | 1261 | |||
XM_005245305.4 | XP_005245362.1 | 7 | 918 | |||
GenBank mRNA | AK022708.1 | - | 3 | 263 | ||
AK297788.1 | - | 5 | 589 | |||
AY017369.1 | - | 7 | 857 | |||
AY302131.1 | - | 5 | 956 | |||
BC025268.1 | - | 2 | 109 | |||
BC109192.1 | - | 7 | 924 | 40001501 | ||
BC109193.1 | - | 7 | 924 | 40001502 |