Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184977.1 | NP_001171906.1 | 4, 5 | 442 | ||
NM_018244.4 | NP_060714.3 | 6, 7 | 646 | |||
NM_199487.2 | NP_955781.2 | 6, 7 | 646 | |||
XM_011528877.1 | XP_011527179.1 | 7, 8 | 593 | |||
XM_011528878.1 | XP_011527180.1 | 5, 6 | 455 | |||
XM_011528879.1 | XP_011527181.1 | 6, 7 | 497 | |||
XM_011528880.1 | XP_011527182.1 | 6, 7 | 555 | |||
XM_011528881.2 | XP_011527183.1 | 5, 6 | 577 | |||
XM_011528882.1 | XP_011527184.1 | 5, 6 | 431 | |||
XM_011528883.1 | XP_011527185.1 | 4, 5 | 357 | |||
XM_011528884.1 | XP_011527186.1 | 4, 5 | 308 | |||
GenBank mRNA | AK001712.1 | - | 6, 7 | 477 | ||
AK022650.1 | - | 6, 7 | 477 | |||
AK022742.1 | - | 6, 7 | 527 | |||
AK225281.1 | - | 6, 7 | 486 | |||
AK225557.1 | - | 6, 7 | 477 | |||
AK293516.1 | - | 4, 5 | 273 | |||
AK295379.1 | - | 4, 5 | 322 | |||
AK308819.1 | - | 6, 7 | 494 | |||
AU280736.1 | - | 6, 7 | 477 | |||
AY050659.1 | - | 6, 7 | 646 | |||
AY050660.1 | - | 4, 5 | 368 | |||
BC105791.1 | - | 6, 7 | 481 | 6458054 | ||
BC119013.1 | - | 6, 7 | 512 | 40085306 | ||
BC122862.1 | - | 6, 7 | 512 | 40085305 | ||
BM919736.1 | - | 6, 7 | 499 |