Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143958.1 | NP_001137430.1 | 5 | 971 | ||
NM_018247.3 | NP_060717.1 | 6 | 1079 | |||
GenBank mRNA | AK001718.1 | - | 6 | 948 | ||
AK292823.1 | - | 5 | 836 | |||
AK293225.1 | - | 7 | 904 | |||
AK295567.1 | - | 7 | 900 | |||
AL162046.1 | - | 3 | 307 | |||
AL832490.1 | - | 6 | 1002 | |||
AL832815.1 | - | 6 | 941 | |||
BC009006.1 | - | 5 | 822 | 4154200 |