Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM30A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143958.1 | NP_001137430.1 | 6 | 2244 | ||
NM_018247.3 | NP_060717.1 | 7 | 2352 | |||
GenBank mRNA | AJ420517.1 | - | 1 | 1025 | ||
AK001718.1 | - | 7 | 2221 | |||
AL162046.1 | - | 4 | 1580 | |||
AL832490.1 | - | 7 | 2275 | |||
AL832815.1 | - | 7 | 2214 | |||
BC009006.1 | - | 6 | 2095 | 4154200 |