Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011649.2 | NP_001011649.1 | 4 | 428 | ||
NM_001272039.1 | NP_001258968.1 | 4 | 428 | |||
NM_018249.5 | NP_060719.4 | 4 | 428 | |||
NR_073554.1 | 4 | 428 | ||||
NR_073555.1 | 4 | 428 | ||||
NR_073556.1 | 4 | 425 | ||||
NR_073557.1 | 4 | 428 | ||||
NR_073558.1 | 4 | 425 | ||||
XM_006717182.1 | XP_006717245.1 | 4 | 417 | |||
XM_006717185.1 | XP_006717248.1 | 4 | 417 | |||
XM_011518860.1 | XP_011517162.1 | 4 | 417 | |||
XM_017014921.1 | XP_016870410.1 | 4 | 417 | |||
XM_017014923.1 | XP_016870412.1 | 4 | 417 | |||
GenBank mRNA | AB046853.2 | - | 4 | 397 | ||
AF448860.1 | - | 4 | 378 | |||
AK122913.1 | - | 4 | 417 | |||
AL833481.1 | - | 4 | 416 | |||
BC019577.1 | - | 4 | 398 | |||
BC136275.1 | - | 4 | 263 | 9020262 | ||
BC143762.1 | - | 4 | 263 | 9052276 | ||
BC146782.1 | - | 4 | 397 | 9007237 | ||
BX537421.1 | - | 4 | 413 | |||
BX537708.1 | - | 4 | 417 | |||
BX537759.1 | - | 4 | 416 | |||
KP294527.1 | - | 4 | 428 |