Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018256.3 | NP_060726.3 | 13 | 2161 | ||
XM_011511469.2 | XP_011509771.1 | 12 | 2038 | |||
GenBank mRNA | AF242546.1 | - | 13 | 1544 | ||
AK001743.1 | - | 13 | 1598 | |||
AK022781.1 | - | 13 | 1606 | |||
AK022782.1 | - | 13 | 1606 | |||
AK056092.1 | - | 13 | 2161 | |||
AK299489.1 | - | 10 | 1131 | |||
BC008082.2 | - | 13 | 1541 | 3050219 | ||
EF011617.1 | - | 13 | 1540 |