Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018263.4 | NP_060733.4 | 2 | 311 | ||
XM_011532950.2 | XP_011531252.1 | 2 | 353 | |||
XM_017004429.1 | XP_016859918.1 | 2 | 141 | |||
GenBank mRNA | AB051472.2 | - | 2 | 311 | ||
AB084279.1 | - | 2 | 174 | |||
BX647527.1 | - | 2 | 384 | |||
BX648218.1 | - | 2 | 336 | |||
CR933613.1 | - | 2 | 355 |