Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018270.5 | NP_060740.1 | 2 | 372 | ||
NR_136405.1 | 2 | 372 | ||||
GenBank mRNA | AB085682.1 | - | 2 | 272 | ||
AK001776.1 | - | 2 | 324 | |||
BC009889.2 | - | 2 | 291 | 4026378 | ||
BC014235.2 | - | 2 | 296 | 4555060 | ||
BC018841.2 | - | 2 | 291 | 3050866 | ||
BM920599.1 | - | 278 |