Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018270.5 | NP_060740.1 | 5 | 1120 | ||
NR_136405.1 | 5 | 1242 | ||||
GenBank mRNA | AB085682.1 | - | 5 | 1020 | ||
AK001776.1 | - | 5 | 1072 | |||
BC009889.2 | - | 5 | 1038 | 4026378 | ||
BC014235.2 | - | 5 | 1043 | 4555060 | ||
BC018841.2 | - | 5 | 1038 | 3050866 |