Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018298.10 | NP_060768.8 | 3 | 468 | ||
XM_005271003.1 | XP_005271060.1 | 4 | 865 | |||
XM_006710750.1 | XP_006710813.1 | 4 | 684 | |||
XM_011541739.2 | XP_011540041.2 | 3 | 392 | |||
XM_011541741.2 | XP_011540043.1 | 3 | 399 | |||
XM_011541742.1 | XP_011540044.1 | 3 | 443 | |||
GenBank mRNA | AF475085.1 | - | 3 | 374 | ||
AK292206.1 | - | 3 | 400 | |||
AK314354.1 | - | 3 | 428 | |||
BC060765.1 | - | 4 | 698 | 30340464 | ||
BP358726.1 | - | 3 | 468 | |||
BX647497.1 | - | 4 | 462 |