Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018339.5 | NP_060809.3 | 2 | 556 | ||
GenBank mRNA | AK002011.1 | - | 2 | 533 | ||
AK315734.1 | - | 2 | 545 | |||
BC007069.1 | - | 2 | 450 | 3997678 | ||
BF979572.1 | - | 551 | ||||
DA154523.1 | - | 2 | 546 |