Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018353.4 | NP_060823.3 | 4 | 1262 | ||
XM_005267833.4 | XP_005267890.1 | 5 | 1309 | |||
XM_017021425.1 | XP_016876914.1 | 6 | 1290 | |||
GenBank mRNA | AB067490.1 | - | 4 | 1263 | ||
AK002048.1 | - | 4 | 1057 | |||
BC051885.1 | - | 4 | 1039 | |||
BC065544.1 | - | 4 | 1053 | 6015439 |