Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168683.1 | NP_001162154.1 | 3 | 374 | ||
NM_018360.2 | NP_060830.2 | 5 | 770 | |||
XM_017029631.1 | XP_016885120.1 | 2 | 744 | |||
GenBank mRNA | AK002071.1 | - | 5 | 765 | ||
AY739713.1 | - | 3 | 324 | |||
BC101572.1 | - | 5 | 737 | 8069078 | ||
BC101576.1 | - | 5 | 737 | 8069082 |