Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300953.1 | NP_001287882.1 | 10 | 1867 | ||
NM_001300954.1 | NP_001287883.1 | 11 | 2049 | |||
NM_001300955.1 | NP_001287884.1 | 10 | 1925 | |||
NM_018367.6 | NP_060837.3 | 11 | 1978 | |||
XM_011545151.2 | XP_011543453.1 | 10 | 1842 | |||
XM_011545152.2 | XP_011543454.1 | 11 | 1914 | |||
GenBank mRNA | AF214454.1 | - | 11 | 1932 | ||
AF327353.1 | - | 11 | 1943 | |||
BC063034.1 | - | 12 | 1938 |