Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018368.3 | NP_060838.3 | 8, 9 | 1048 | ||
XM_011535941.1 | XP_011534243.1 | 8, 9 | 898 | |||
GenBank mRNA | AF113224.1 | - | 8, 9 | 768 | ||
AF208863.1 | - | 7, 8 | 896 | |||
AF211480.1 | - | 5, 6 | 386 | |||
AK002102.1 | - | 8, 9 | 797 | |||
AK225187.1 | - | 8, 9 | 797 | |||
AK225537.1 | - | 8, 9 | 782 | |||
AY136817.1 | - | 7, 8 | 553 | |||
BC010360.1 | - | 6, 7 | 614 | |||
BC047073.1 | - | 8, 9 | 887 | 5274221 | ||
BX647737.1 | - | 7, 8 | 640 |