Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018368.3 | NP_060838.3 | 9 | 1108 | ||
XM_011535941.1 | XP_011534243.1 | 9 | 958 | |||
GenBank mRNA | AF113224.1 | - | 9 | 828 | ||
AF208863.1 | - | 8 | 956 | |||
AF211480.1 | - | 6 | 446 | |||
AK002102.1 | - | 9 | 857 | |||
AK225187.1 | - | 9 | 857 | |||
AK225537.1 | - | 9 | 842 | |||
AK290069.1 | - | 9 | 988 | |||
AY136817.1 | - | 8 | 613 | |||
BC010360.1 | - | 7 | 674 | |||
BC047073.1 | - | 9 | 947 | 5274221 | ||
BX647737.1 | - | 8 | 700 |