Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNJ2BP-COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202547.2 | NP_001189476.1 | 3 | 384 | ||
NM_001202548.2 | NP_001189477.1 | 3 | 384 | |||
NM_001202549.2 | NP_001189478.1 | 3 | 384 | |||
GenBank mRNA | FY210787.1 | - | 3 | 278 | ||
FY211163.1 | - | 2 | 141 | |||
FY213292.1 | - | 3 | 278 | |||
FY213836.1 | - | 3 | 278 |
Gene Symbol: SYNJ2BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018373.2 | NP_060843.2 | 3 | 384 | ||
GenBank mRNA | AK002133.1 | - | 3 | 339 | ||
AK311719.1 | - | 2 | 218 | |||
AY534623.1 | - | 3 | 353 | |||
BC007704.2 | - | 3 | 286 | 3952366 | ||
BP195583.1 | - | 3 | 384 |