Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018434.5 | NP_060904.2 | 8 | 1649 | ||
GenBank mRNA | AF155650.1 | - | 7 | 948 | ||
AK313089.1 | - | 8 | 1132 | |||
AY083998.1 | - | 8 | 1631 | |||
BC017100.1 | - | 8 | 1231 | |||
BC065244.1 | - | 7 | 894 | |||
BC082267.1 | - | 8 | 1246 | |||
BC108306.1 | - | 8 | 1644 | 6137501 | ||
BC113864.1 | - | 9 | 1079 | 40033418 | ||
BC114472.1 | - | 8 | 1011 | |||
DB572650.2 | - | 1 | 75 |