Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEMGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018437.4 | NP_060907.2 | 5 | 1707 | ||
NM_197978.2 | NP_932095.1 | 4 | 1761 | |||
XM_005252086.1 | XP_005252143.1 | 5 | 1669 | |||
XM_011518845.2 | XP_011517147.1 | 5 | 1581 | |||
XM_011518846.2 | XP_011517148.1 | 5 | 1691 | |||
GenBank mRNA | AF116617.1 | - | 2 | 1163 | ||
AF130060.2 | - | 4 | 1494 | |||
AF228713.2 | - | 5 | 1614 | |||
AF306681.1 | - | 2 | 585 | |||
AF306682.1 | - | 4 | 1543 | |||
AL833094.1 | - | 4 | 1553 | |||
AY244805.1 | - | 4 | 1707 | |||
AY255672.1 | - | 4 | 1612 |