Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018474.4 | NP_060944.3 | 3 | 357 | ||
XM_011529296.2 | XP_011527598.1 | 3 | 360 | |||
XM_011529297.2 | XP_011527599.1 | 3 | 349 | |||
XM_011529298.2 | XP_011527600.1 | 3 | 342 | |||
XM_017027951.1 | XP_016883440.1 | 3 | 259 | |||
XM_017027953.1 | XP_016883442.1 | 3 | 331 | |||
GenBank mRNA | AF451990.1 | - | 3 | 335 | ||
AK025476.1 | - | 3 | 288 | |||
BC039296.1 | - | 3 | 319 | |||
BC105093.1 | - | 3 | 260 | 8144096 | ||
BC113370.1 | - | 3 | 260 | 8322422 |