Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163022.1 | NP_001156494.1 | 4 | 841 | ||
NM_001163023.1 | NP_001156495.1 | 3 | 751 | |||
NM_001276389.1 | NP_001263318.1 | 3 | 778 | |||
NM_018474.4 | NP_060944.3 | 5 | 1004 | |||
XM_011529296.2 | XP_011527598.1 | 5 | 1007 | |||
XM_011529297.2 | XP_011527599.1 | 5 | 996 | |||
XM_011529298.2 | XP_011527600.1 | 5 | 989 | |||
XM_011529299.2 | XP_011527601.1 | 4 | 836 | |||
XM_017027951.1 | XP_016883440.1 | 6 | 950 | |||
XM_017027952.1 | XP_016883441.1 | 4 | 836 | |||
XM_017027953.1 | XP_016883442.1 | 6 | 1022 | |||
XM_017027954.1 | XP_016883443.1 | 4 | 836 | |||
GenBank mRNA | AF220187.1 | - | 1 | 36 | ||
AF451990.1 | - | 5 | 981 | |||
AK025476.1 | - | 5 | 935 | |||
AK293166.1 | - | 3 | 682 | |||
AK293567.1 | - | 3 | 722 | |||
AK295355.1 | - | 4 | 740 | |||
AK296399.1 | - | 1 | 270 | |||
AL110196.1 | - | 1 | 283 | |||
BC039296.1 | - | 5 | 965 | |||
BC045826.1 | - | 4 | 728 | 5262165 | ||
BC065550.1 | - | 3 | 585 | |||
BC090879.2 | - | 1 | 409 | |||
BC105093.1 | - | 5 | 907 | 8144096 | ||
BC113370.1 | - | 5 | 907 | 8322422 |