Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 7477 | ||
NM_005197.3 | NP_005188.2 | 6 | 7528 | |||
GenBank mRNA | AK055175.1 | - | 2 | 3145 | ||
AK123929.1 | - | 1 | 2153 | |||
BC010460.1 | - | 1 | 1269 |