Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018665.2 | NP_061135.2 | 1 | 475 | ||
XM_011535926.1 | XP_011534228.1 | 1 | 475 | |||
XM_011535928.2 | XP_011534230.1 | 1 | 177 | |||
XM_017011003.1 | XP_016866492.1 | 1 | 143 | |||
GenBank mRNA | AJ278110.1 | - | 1 | 338 | ||
AK303324.1 | - | 1 | 184 | |||
AL136751.1 | - | 1 | 163 | |||
BC066938.1 | - | 1 | 156 | 4829072 | ||
BX646425.1 | - | 1 | 222 |