Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 2 | 812 | ||
GenBank mRNA | AB035966.1 | - | 2 | 289 | ||
AF353942.1 | - | 2 | 289 | |||
AJ278245.1 | - | 2 | 420 | |||
AK095857.1 | - | 2 | 812 | |||
AK301747.1 | - | 2 | 218 | |||
AK307204.1 | - | 3 | 573 | |||
AK313326.1 | - | 2 | 557 | |||
AL713665.2 | - | 2 | 204 | |||
BC070049.1 | - | 2 | 508 | 30343188 |