Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018708.2 | NP_061178.1 | 1 | 1063 | ||
GenBank mRNA | AJ311359.1 | - | 1 | 923 | ||
AK074232.1 | - | 1 | 1021 | |||
AK126532.1 | - | 2 | 791 | |||
AK315554.1 | - | 1 | 1035 | |||
AY356352.1 | - | 1 | 1017 | |||
BC004988.1 | - | 1 | 1021 | 2957950 |