Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003518.3 | NP_003509.1 | 1 | 141 | ||
GenBank mRNA | AK311935.1 | - | 1 | 190 | ||
BC082232.1 | - | 1 | 168 |