Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018903.3 | NP_061726.1 | 1 | 2510 | ||
NM_031864.2 | NP_114070.1 | 1 | 2510 | |||
GenBank mRNA | AF152308.1 | - | 1 | 2339 | ||
AF152477.1 | - | 1 | 2339 | |||
AK075556.1 | - | 1 | 1391 | |||
BC112280.1 | - | 1 | 2371 | 8327748 | ||
BC113372.1 | - | 1 | 2371 | 8322424 |