Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018908.2 | NP_061731.1 | 1 | 629 | ||
NM_031501.1 | NP_113689.1 | 1 | 629 | |||
GenBank mRNA | AF152313.1 | - | 1 | 629 | ||
AF152483.1 | - | 1 | 629 | |||
BC033735.1 | - | 1 | 768 | 5175471 |