Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018919.2 | NP_061742.1 | 1 | 999 | ||
NM_032086.1 | NP_114475.1 | 1 | 999 | |||
GenBank mRNA | AB209178.1 | - | 1 | 1169 | ||
AF152326.1 | - | 1 | 999 | |||
AF152513.1 | - | 1 | 999 | |||
BC109240.1 | - | 1 | 199 | |||
BC136702.1 | - | 1 | 1090 | 9020692 | ||
BC136703.1 | - | 1 | 1090 | 9020693 | ||
BC146651.1 | - | 1 | 1158 | 40147340 |