Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003531.2 | NP_003522.1 | 1 | 412 | ||
GenBank mRNA | BC127610.1 | - | 1 | 412 | 40002514 |